Bis(ethylcyclopentadienyl)nickel(II), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(ethylcyclopentadienyl)nickel

M-Clarity™ Qualitätsstufe = MQ100

510483-5G
SIAL510483-5GEA 213 EUR
SIAL510483-5G
Bis(ethylcyclopentadienyl)nickel(II), Sigma-Aldrich®
Bis(ethylcyclopentadienyl)nickel(II)
Formel: C₁₄H₁₈Ni
Molecular Weight: 244,986 g/mol
MDL: MFCD01862458
CAS-Nummer: 31886-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR