Bis(ethylcyclopentadienyl)cobalt(II), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

510645-1G
SIAL510645-1GEA 195 EUR
SIAL510645-1G
Bis(ethylcyclopentadienyl)cobalt(II), Sigma-Aldrich®
Bis(ethylcyclopentadienyl)cobalt(II)
Formel: C₁₄H₁₈Co
MDL: MFCD01863655
CAS-Nummer: 55940-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR