6-Chlornicotinonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

510734-5G
SIAL510734-5GEA 33.7 EUR
SIAL510734-5G
6-Chlornicotinonitril, Sigma-Aldrich®
6-Chlornicotinonitril
Formel: C₆H₃ClN₂
MDL: MFCD00084941
CAS-Nummer: 33252-28-7
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR