Aluminiumtri-sec-butanolat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide

511609-5G
SIAL511609-5GEA 185 EUR
SIAL511609-5G
Aluminiumtri-sec-butanolat, Sigma-Aldrich®
Aluminiumtri-sec-butanolat
Formel: C₁₂H₂₇AlO₃
Molecular Weight: 246,33 g/mol
MDL: MFCD00009327
CAS-Nummer: 2269-22-9
EINECS: 218-871-2
UN: 1992
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR