Natrium-2-ethylhexanoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Ethylhexansäure Natriumsalz, 2-Ethylcapronsäure Natriumsalz, Heptan-3-carbonsäure Natriumsalz
M-Clarity™ Qualitätsstufe = MQ200
511692-1KG
SIAL511692-1KGEA
164
EUR
SIAL511692-1KG
Natrium-2-ethylhexanoat, Sigma-Aldrich®
Natrium-2-ethylhexanoat
Formel:
C₈H₁₅NaO₂ Molecular Weight: 166,2 g/mol |
MDL:
MFCD00014007 CAS-Nummer: 19766-89-3 EINECS: 243-283-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...