Methyl-4-isocyanatobenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-isocyanatobenzoate

M-Clarity™ Qualitätsstufe = MQ100

511935-5G
SIAL511935-5GEA 202 EUR
SIAL511935-5G
Methyl-4-isocyanatobenzoat, Sigma-Aldrich®
Methyl-4-isocyanatobenzoat
Formel: C₉H₇NO₃
Molecular Weight: 177,1568 g/mol
MDL: MFCD01863690
CAS-Nummer: 23138-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR