3-Benzo[b]thiophenboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzothiophene-3-boronic acid, Benzo[b]thiophen-3-ylboronic acid

M-Clarity™ Qualitätsstufe = MQ100

512117-5G
SIAL512117-5GEA 125 EUR
SIAL512117-5G
3-Benzo[b]thiophenboronsäure, Sigma-Aldrich®
3-Benzo[b]thiophenboronsäure
Formel: C₈H₇BO₂S
Molecular Weight: 178.02 g/mol
MDL: MFCD01114669
CAS-Nummer: 113893-08-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR