Furan-3-boronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Furanboronic acid, 3-Furanylboronic acid

512168-1G
SIAL512168-1GEA 87.4 EUR
SIAL512168-1G
Furan-3-boronsäure, Sigma-Aldrich®
Furan-3-boronsäure
Formel: C₄H₅BO₃
Molecular Weight: 111,89 g/mol
MDL: MFCD01319007
CAS-Nummer: 55552-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR