3-Chlor-4-fluorphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dihydroxy(3-chlor-4-fluorphenyl)boran, (3-Chlor-4-fluorphenyl)metaborsäure, 3-Chlor-4-fluorbenzolboronsäure

M-Clarity™ Qualitätsstufe = MQ100

512230-5G
SIAL512230-5GEA 53 EUR
SIAL512230-5G
3-Chlor-4-fluorphenylboronsäure, Sigma-Aldrich®
3-Chlor-4-fluorphenylboronsäure
Formel: C₆H₅BClFO₂
Molecular Weight: 174,37 g/mol
MDL: MFCD00051800
CAS-Nummer: 144432-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR