4-(Hydroxymethyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Hydroxymethyl)benzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

512338-10G 512338-1G
SIAL512338-10GEA 248 EUR
SIAL512338-10G SIAL512338-1G
4-(Hydroxymethyl)phenylboronsäure, Sigma-Aldrich®
4-(Hydroxymethyl)phenylboronsäure
Formel: C₇H₉BO₃
MDL: MFCD00792672
CAS-Nummer: 59016-93-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR