4-(Benzyloxy)benzoesäure, Sigma-Aldrich®

Lieferant: Merck

512354-5G
SIAL512354-5GEA 35.2 EUR
SIAL512354-5G
4-(Benzyloxy)benzoesäure, Sigma-Aldrich®
4-(Benzyloxy)benzoesäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
MDL: MFCD00016527
CAS-Nummer: 1486-51-7
EINECS: 216-066-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR