3-Chlor-L-tyrosin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-Tyr(3-Cl)-OH

M-Clarity™ Qualitätsstufe = MQ100

512443-1G
SIAL512443-1GEA 84.9 EUR
SIAL512443-1G
3-Chlor-L-tyrosin, Sigma-Aldrich®
3-Chlor-L-tyrosin
Formel: C₉H₁₀ClNO₃
Molecular Weight: 215,7 g/mol
MDL: MFCD00045865
CAS-Nummer: 7423-93-0
EINECS: 231-050-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR