(R)-(+)-Indolin-2-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

51266-500MG-F
SIAL51266-500MG-FEA 76.1 EUR
SIAL51266-500MG-F
(R)-(+)-Indolin-2-carbonsäure, Sigma-Aldrich®
(R)-(+)-Indolin-2-carbonsäure
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
MDL: MFCD00792496
CAS-Nummer: 98167-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR