5-Brom-2-methoxyphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-2-methoxybenzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

512702-5G
SIAL512702-5GEA 87.3 EUR
SIAL512702-5G
5-Brom-2-methoxyphenylboronsäure, Sigma-Aldrich®
5-Brom-2-methoxyphenylboronsäure
Formel: C₇H₈BBrO₃
Molecular Weight: 230,85 g/mol
MDL: MFCD00093409
CAS-Nummer: 89694-45-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR