3-Formyl-4-methoxyphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Formyl-4-methoxybenzeneboronic acid

512869-5G
SIAL512869-5GEA 79 EUR
SIAL512869-5G
3-Formyl-4-methoxyphenylboronsäure, Sigma-Aldrich®
3-Formyl-4-methoxyphenylboronsäure
Formel: C₈H₉BO₄
MDL: MFCD02093661
CAS-Nummer: 121124-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR