5,5'-Dimethyl-2,2'-bipyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline

M-Clarity™ Qualitätsstufe = MQ100

513040-25G 513040-5G
SIAL513040-25GEA 309 EUR
SIAL513040-25G SIAL513040-5G
5,5'-Dimethyl-2,2'-bipyridin, Sigma-Aldrich®
5,5'-Dimethyl-2,2'-bipyridyl
Formel: C₁₂H₁₂N₂
MDL: MFCD01740554
CAS-Nummer: 1762-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR