5,5'-Dimethyl-2,2'-bipyridin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline
M-Clarity™ Qualitätsstufe = MQ100
513040-25G
513040-5G
SIAL513040-25GEA
309
EUR
SIAL513040-25G
SIAL513040-5G
5,5'-Dimethyl-2,2'-bipyridin, Sigma-Aldrich®
5,5'-Dimethyl-2,2'-bipyridyl
Formel:
C₁₂H₁₂N₂ |
MDL:
MFCD01740554 CAS-Nummer: 1762-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...