2-Iodpyrazin, Sigma-Aldrich®

Lieferant: Merck

513164-5G
SIAL513164-5GEA 166 EUR
SIAL513164-5G
2-Iodpyrazin, Sigma-Aldrich®
2-Iodopyrazine
Formel: C₄H₃IN₂
Molecular Weight: 205.99 g/mol
MDL: MFCD01319019
CAS-Nummer: 32111-21-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR