4-Carboxyphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

M-Clarity™ Qualitätsstufe = MQ100

513490-5G
SIAL513490-5GEA 29.2 EUR
SIAL513490-5G
4-Carboxyphenylboronsäurepinakolester, Sigma-Aldrich®
4-Carboxyphenylboronsäurepinakolester
Formel: C₁₃H₁₇BO₄
Molecular Weight: 248.08 g/mol
MDL: MFCD01863710
CAS-Nummer: 180516-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR