5-Indolcarbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indol-5-carboxaldehyd

M-Clarity™ Qualitätsstufe = MQ100

513830-5G
SIAL513830-5GEA 43.2 EUR
SIAL513830-5G
5-Indolcarbaldehyd, Sigma-Aldrich®
5-Indolcarbaldehyd
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
MDL: MFCD02093664
CAS-Nummer: 1196-69-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR