1,2,4-Trichlor-5-iodbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4,5-Trichloriodbenzol, 2,4,5-Trichlorjodbenzol, 1-Iod-2,4,5-trichlorbenzol

M-Clarity™ Qualitätsstufe = MQ100

513873-5G
SIAL513873-5GEA 74.9 EUR
SIAL513873-5G
1,2,4-Trichlor-5-iodbenzol, Sigma-Aldrich®
1,2,4-Trichlor-5-iodbenzol
Formel: C₆H₂Cl₃I
Molecular Weight: 307,34 g/mol
MDL: MFCD00051450
CAS-Nummer: 7145-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR