2,3-Dichlorphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

514047-25G
SIAL514047-25GEA 182 EUR
SIAL514047-25G
2,3-Dichlorphenylboronsäure, Sigma-Aldrich®
2,3-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190.82 g/mol
MDL: MFCD01075703
CAS-Nummer: 151169-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR