Ytterbium(III)isopropoxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ytterbium(III) i-propoxide

M-Clarity™ Qualitätsstufe = MQ100

514063-5G
SIAL514063-5GEA 594 EUR
SIAL514063-5G
Ytterbium(III)isopropoxid, Sigma-Aldrich®
Ytterbium(III)isopropoxid
Formel: C₉H₂₁O₃Yb
Molecular Weight: 350,30 g/mol
MDL: MFCD00145532
CAS-Nummer: 6742-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR