(S)-(-)-α-Ethylbenzylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(-)-1-Phenylpropylamine

51432-5G-F
SIAL51432-5G-FEA 71 EUR
SIAL51432-5G-F
(S)-(-)-α-Ethylbenzylamin, Sigma-Aldrich®
(S)-(-)-α-Ethylbenzylamin
Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
MDL: MFCD00082356
CAS-Nummer: 3789-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR