2,2-Dibutyl-1,3-propandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5,5-Bis(hydroxymethyl)nonan

M-Clarity™ Qualitätsstufe = MQ100

514454-25G
SIAL514454-25GEA 102 EUR
SIAL514454-25G
2,2-Dibutyl-1,3-propandiol, Sigma-Aldrich®
2,2-Dibutyl-1,3-propandiol
Formel: C₁₁H₂₄O₂
Molecular Weight: 188,31 g/mol
MDL: MFCD01863725
CAS-Nummer: 24765-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR