4-Methoxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Methoxy-TEMPO), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy

M-Clarity™ Qualitätsstufe = MQ200

514586-5G
SIAL514586-5GEA 154 EUR
SIAL514586-5G
4-Methoxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Methoxy-TEMPO), Sigma-Aldrich®
4-Methoxy-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Methoxy-TEMPO)
Formel: C₁₀H₂₀NO₂
Molecular Weight: 186,27 g/mol
MDL: MFCD00270334
CAS-Nummer: 95407-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR