4-Brom-4'-methoxybiphenyl, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Bromo-4'-methoxy-1,1'-biphenyl, 1-Bromo-4-(4-methoxyphenyl)benzene

M-Clarity™ Qualitätsstufe = MQ100

514616-5G
SIAL514616-5GEA 279 EUR
SIAL514616-5G
4-Brom-4'-methoxybiphenyl, Sigma-Aldrich®
4-Brom-4'-methoxybiphenyl
Formel: C₁₃H₁₁BrO
Molecular Weight: 263,1298 g/mol
MDL: MFCD00671954
CAS-Nummer: 58743-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR