2,2,2,3',4'-Pentafluoracetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

514624-1G
SIAL514624-1GEA 265 EUR
SIAL514624-1G
2,2,2,3',4'-Pentafluoracetophenon, Sigma-Aldrich®
2,2,2,3',4'-Pentafluoracetophenon
Formel: C₈H₃F₅O
MDL: MFCD01319992
CAS-Nummer: 302912-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR