4-Nitroindan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Nitro-2,3-dihydro-1H-indene

M-Clarity™ Qualitätsstufe = MQ100

515035-5G
SIAL515035-5GEA 89.5 EUR
SIAL515035-5G
4-Nitroindan, Sigma-Aldrich®
4-Nitroindan
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
MDL: MFCD00078173
CAS-Nummer: 34701-14-9
EINECS: 252-159-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR