1-Brom-3-isopropoxybenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromophenyl isopropyl ether

M-Clarity™ Qualitätsstufe = MQ200

515507-5G
SIAL515507-5GEA 42 EUR
SIAL515507-5G
1-Brom-3-isopropoxybenzol, Sigma-Aldrich®
1-Brom-3-isopropoxybenzol
Formel: C₉H₁₁BrO
Molecular Weight: 215,09 g/mol
MDL: MFCD00070756
CAS-Nummer: 131738-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR