Isoindolin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

515574-5G
SIAL515574-5GEA 327 EUR
SIAL515574-5G
Isoindolin, Sigma-Aldrich®
Isoindolin
Formel: C₈H₉N
MDL: MFCD00605324
CAS-Nummer: 496-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR