Nitrosiumhexafluoroantimonat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Nitrosylhexafluoroantimonat
M-Clarity™ Qualitätsstufe = MQ100
515930-5G
SIAL515930-5GEA
138
EUR
SIAL515930-5G
Nitrosiumhexafluoroantimonat, Sigma-Aldrich®
Nitrosiumhexafluoroantimonat
Formel:
F₆NOSb Molecular Weight: 265,76 g/mol |
MDL:
MFCD00042550 CAS-Nummer: 16941-06-3 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...