4-Chlor-2-iodanilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

516139-5G
SIAL516139-5GEA 116 EUR
SIAL516139-5G
4-Chlor-2-iodanilin, Sigma-Aldrich®
4-Chlor-2-iodanilin
Formel: C₆H₅ClIN
MDL: MFCD01863737
CAS-Nummer: 63069-48-7
EINECS: 263-839-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR