2,4-Diethoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

516309-5G
SIAL516309-5GEA 318 EUR
SIAL516309-5G
2,4-Diethoxybenzaldehyd, Sigma-Aldrich®
2,4-Diethoxybenzaldehyde
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,2271 g/mol
MDL: MFCD00995010
CAS-Nummer: 22924-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR