4-Methoxy-3-nitrobenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

516422-5G
SIAL516422-5GEA 47.9 EUR
SIAL516422-5G
4-Methoxy-3-nitrobenzaldehyd, Sigma-Aldrich®
4-Methoxy-3-nitrobenzaldehyde
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
MDL: MFCD00126498
CAS-Nummer: 31680-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR