2-Chlor-4-fluoranisol, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

516740-5G
SIAL516740-5GEA 22.3 EUR
SIAL516740-5G
2-Chlor-4-fluoranisol, Sigma-Aldrich®
2-Chlor-4-fluoranisol
Formel: C₇H₆ClFO
Molecular Weight: 160,58 g/mol
MDL: MFCD00672965
CAS-Nummer: 2267-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR