5-Brom-2-chloranisol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

516767-5G
SIAL516767-5GEA 61.6 EUR
SIAL516767-5G
5-Brom-2-chloranisol, Sigma-Aldrich®
5-Brom-2-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,48 g/mol
MDL: MFCD00078659
CAS-Nummer: 16817-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR