Iridium(IV)chlorid Hydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

516996-1G
SIAL516996-1GEA 400 EUR
SIAL516996-1G
Iridium(IV)chlorid Hydrat, Sigma-Aldrich®
Iridium(IV)chlorid Hydrat
Formel: Cl₄Ir·xH₂O
Molecular Weight: 334,03 g/mol (anhy)
MDL: MFCD00150243
CAS-Nummer: 207399-11-9
EINECS: 233-048-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR