3,5-Dimethylbenzaldehyd, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
518050-5G
SIAL518050-5GEA
45.5
EUR
SIAL518050-5G
3,5-Dimethylbenzaldehyd, Sigma-Aldrich®
3,5-Dimethylbenzaldehyde
Formel:
C₉H₁₀O Molecular Weight: 134,18 g/mol Dichte: 1,09 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082777 CAS-Nummer: 5779-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...