1-Boc-indol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-Butyl 1-indolecarboxylate

518107-25ML
SIAL518107-25MLEA 180 EUR
SIAL518107-25ML
1-Boc-indol, Sigma-Aldrich®
1-Boc-indol
Formel: C₁₃H₁₅NO₂
Molecular Weight: 217,27 g/mol
MDL: MFCD02093939
CAS-Nummer: 75400-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR