4-Fluor-3-nitrobenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

518301-1G
SIAL518301-1GEA 26 EUR
SIAL518301-1G
4-Fluor-3-nitrobenzaldehyd, Sigma-Aldrich®
4-Fluoro-3-nitrobenzaldehyde
Formel: C₇H₄FNO₃
Molecular Weight: 169,11 g/mol
MDL: MFCD01861388
CAS-Nummer: 42564-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR