3-Chlor-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

518409-5G
SIAL518409-5GEA 106 EUR
SIAL518409-5G
3-Chlor-o-toluylsäure, Sigma-Aldrich®
3-Chlor-o-toluylsäure
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
MDL: MFCD00053312
CAS-Nummer: 7499-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR