4-Amino-2-chlor-6,7-dimethoxychinazolin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline
M-Clarity™ Qualitätsstufe = MQ100
518654-25G
SIAL518654-25GEA
72.3
EUR
SIAL518654-25G
4-Amino-2-chlor-6,7-dimethoxychinazolin, Sigma-Aldrich®
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Formel:
C₁₀H₁₀ClN₃O₂ Molecular Weight: 239,66 g/mol |
MDL:
MFCD00051734 CAS-Nummer: 23680-84-4 EINECS: 245-821-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...