2-Amino-5-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

518689-1G
SIAL518689-1GEA 50.4 EUR
SIAL518689-1G
2-Amino-5-fluorpyridin, Sigma-Aldrich®
2-Amino-5-fluorpyridin
Formel: C₅H₅FN₂
MDL: MFCD01861120
CAS-Nummer: 21717-96-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR