4-(Trifluormethyl)nicotinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

518697-1G
SIAL518697-1GEA 53.3 EUR
SIAL518697-1G
4-(Trifluormethyl)nicotinsäure, Sigma-Aldrich®
4-(Trifluormethyl)nicotinsäure
Formel: C₇H₄F₃NO₂
Molecular Weight: 191,11 g/mol
MDL: MFCD00082626
CAS-Nummer: 158063-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR