3,5-Dimethyl-4-hydroxyphenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,5-Dimethyl-4-hydroxyphenylboronic acid pinacol ester, 3,5-Dimethy-4-hydroxyphenylboronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

518794-5G
SIAL518794-5GEA 109 EUR
SIAL518794-5G
3,5-Dimethyl-4-hydroxyphenylboronsäurepinakolester, Sigma-Aldrich®
3,5-Dimethyl-4-hydroxyphenylboronsäurepinakolester
Formel: C₁₄H₂₁BO₃
Molecular Weight: 248.13 g/mol
MDL: MFCD02093724
CAS-Nummer: 269410-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR