1-Fluor-3-ethinylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethinyl-3-fluorbenzol, (3-Fluorphenyl)acetylen, (3-Fluorphenyl)ethin

519405-5G
SIAL519405-5GEA 63.8 EUR
SIAL519405-5G
1-Fluor-3-ethinylbenzol, Sigma-Aldrich®
1-Fluor-3-ethinylbenzol
Formel: C₈H₅F
Molecular Weight: 120.13 g/mol
MDL: MFCD03093798
CAS-Nummer: 2561-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR