3-Ethinylanisol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methoxy-3-ethinylbenzol, 1-Ethinyl-3-methoxybenzol, (3-Methoxyphenyl)acetylen, (3-Methoxyphenyl)ethin

M-Clarity™ Qualitätsstufe = MQ100

519413-5G
SIAL519413-5GEA 243 EUR
SIAL519413-5G
3-Ethinylanisol, Sigma-Aldrich®
3-Ethinylanisol
Formel: C₉H₈O
Molecular Weight: 132,16 g/mol
MDL: MFCD00160810
CAS-Nummer: 768-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR