5-Brom-2-nitropyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

520411-5G
SIAL520411-5GEA 51.3 EUR
SIAL520411-5G
5-Brom-2-nitropyridin, Sigma-Aldrich®
5-Brom-2-nitropyridin
Formel: C₅H₃BrN₂O₂
Molecular Weight: 203 g/mol
MDL: MFCD00160411
CAS-Nummer: 39856-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR