2-Brom-6-methoxypyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

520535-5ML
SIAL520535-5MLEA 73.3 EUR
SIAL520535-5ML
2-Brom-6-methoxypyridin, Sigma-Aldrich®
2-Bromo-6-methoxypyridine
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD00088345
CAS-Nummer: 40473-07-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR